missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Amino-N-Boc-L-proline methyl ester hydrochloride, 97%

Product Code. p-7034866
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15459905 1 g 1g
15469905 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15459905 Supplier Thermo Scientific Alfa Aesar Supplier No. H52566.03
Add to basket
This item is not returnable. View return policy

CAS: 334999-32-5 | C11H21ClN2O4 | 280.75 g/mol

trans-4-Amino-N-Boc-L-proline methyl ester hydrochloride acts as a pharmaceutical intermediate. It is used to prepare N-Boc-protected proline methyl ester of calix[4]arene, which is efficiently catalyzes the enantioselective aldol reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-Amino-N-Boc-L-proline methyl ester hydrochloride acts as a pharmaceutical intermediate. It is used to prepare N-Boc-protected proline methyl ester of calix[4]arene, which is efficiently catalyzes the enantioselective aldol reaction.

Notes
Store in a cool place. Moisture sensitive. Incompatible with oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 334999-32-5
Molecular Formula C11H21ClN2O4
Molecular Weight (g/mol) 280.75
MDL Number MFCD03787929
InChI Key KFYCQKLSGMAVQH-WLYNEOFISA-N
Synonym 2s,4r-1-tert-butyl 2-methyl 4-aminopyrrolidine-1,2-dicarboxylate hydrochloride, n-boc-trans-4-amino-l-proline methyl ester hydrochloride, 2s,4r-1-boc-4-amino-l-proline methyl ester hcl, 2s,4r-4-amino-1-boc-pyrrolidine-2-carboxylic acid methyl ester-hcl, 1-tert-butyl 2-methyl 2s,4r-4-aminopyrrolidine-1,2-dicarboxylate hydrochloride, n-boc-trans-4-amino-l-proline methyl ester hydrochloride salt, methyl trans-3-amino-1-boc-5-pyrrolidinecarboxylate hcl, boc-trans-4-amino-l-proline methyl ester hydrochloride salt, 2s,4r-1-boc-4-amino-l-proline methyl ester hydrochloride, n-boc-trans-4-amino-l-proline methyl ester, hydrochloride salt
PubChem CID 17750455
SMILES [H+].[Cl-].COC(=O)[C@@H]1C[C@@H](N)CN1C(=O)OC(C)(C)C

Specifications

Quantity 1 g
IUPAC Name hydrogen 1-tert-butyl 2-methyl (2S,4R)-4-aminopyrrolidine-1,2-dicarboxylate chloride
Formula Weight 280.75
Percent Purity 97%
Chemical Name or Material trans-4-Amino-N-Boc-L-proline methyl ester hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.