missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ trans-4-(4-n-Heptylcyclohexyl)benzonitrile, 97%, Thermo Scientific™

Product Code. 15401668
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as intermediates of liquid crystals.

Solubility
Slightly soluble in water.

Notes
Storage temperature: 2 - 4°Cand Keep container tightly closed in a dryand well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61204-03-3
Molecular Formula C20H29N
Molecular Weight (g/mol) 283.459
MDL Number MFCD01941020
InChI Key NSGMZTNTQKRAFA-UHFFFAOYSA-N
Synonym 4-trans-4-heptylcyclohexyl benzonitrile, 4-4-heptylcyclohexyl benzonitrile, trans-4-4-heptylcyclohexyl benzonitrile, trans-4-heptyl 4-cyanophenyl cyclohexane, benzonitrile, 4-trans-4-heptylcyclohexyl, p-4-heptylcyclohexyl benzonitrile, 4-1s,4r-4-heptylcyclohexyl benzonitrile, trans-4-4-heptylcyclohexylbenzonitrile, pch-7, nsgmztntqkrafa-uhfffaoysa
PubChem CID 109064
IUPAC Name 4-(4-heptylcyclohexyl)benzonitrile
SMILES CCCCCCCC1CCC(CC1)C2=CC=C(C=C2)C#N

Specifications

Melting Point 33°C
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 283.46
Percent Purity 97%
Chemical Name or Material trans-4-(4-n-Heptylcyclohexyl)benzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'