missing translation for 'onlineSavingsMsg'
Learn More

trans-4-(4-n-Heptylcyclohexyl)benzoic acid, 98%, Thermo Scientific™

Product Code. 15448636
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101532-38-1
Molecular Formula C19H29Br
Molecular Weight (g/mol) 337.345
MDL Number MFCD09838992
InChI Key OKIVMWBXCXBOGL-UHFFFAOYSA-N
Synonym 1-bromo-4-trans-4-heptylcyclohexyl benzene, 1-bromo-4-4-heptylcyclohexyl benzene, 1-bromo-4-1s,4r-4-heptylcyclohexyl benzene
PubChem CID 15135337
IUPAC Name 1-bromo-4-(4-heptylcyclohexyl)benzene
SMILES CCCCCCCC1CCC(CC1)C2=CC=C(C=C2)Br

Specifications

Quantity 250 mg
Beilstein 4748130
Formula Weight 302.46
Percent Purity 98%
Physical Form Solid
Chemical Name or Material trans-4-(4-n-Heptylcyclohexyl)benzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'