missing translation for 'onlineSavingsMsg'
Learn More

trans-3'-Hydroxy Cotinine, TRC

Product Code. 30407353
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30407353 1 mg 1mg
30397876 25 mg 25mg
30390707 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30407353 Supplier Toronto Research Chemicals Supplier No. TRCH9245001MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trans-3'-Hydroxycotinine
CAS 34834-67-8
Purity Grade Notes HPLC
Molecular Formula C10 H12 N2 O2
Quantity 1 mg
Synonym 2-Pyrrolidinone, 3-hydroxy-1-methyl-5-(3-pyridinyl)-, (3R,5S)-, 2-Pyrrolidinone, 3-hydroxy-1-methyl-5-(3-pyridinyl)-, (3R-trans)-, Cotinine, hydroxy- (6CI), (3R,5S)-3-Hydroxy-1-methyl-5-(3-pyridinyl)-2-pyrrolidinone, (3S-trans)-3-Hydroxycotinine, (3'R,5'S)-3'-Hydroxycotinine, Hydroxycotinine, trans-1-Methyl-3(R)-hydroxy-5(S)-3-pyridyl-2-pyrrolidinone, trans-3'-Hydroxycotinine
InChI Formula InChI=1S/C10H12N2O2/c1-12-8(5-9(13)10(12)14)7-3-2-4-11-6-7/h2-4,6,8-9,13H,5H2,1H3/t8-,9+/m0/s1
SMILES CN1[C@@H](C[C@@H](O)C1=O)c2cccnc2
IUPAC Name (3R,5S)-3-hydroxy-1-methyl-5-pyridin-3-ylpyrrolidin-2-one
Molecular Weight (g/mol) 192.21
Formula Weight 192.0899
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.