missing translation for 'onlineSavingsMsg'
Learn More

trans-3'-Hydroxy Cotinine-d3 Acetate, TRC

Product Code. 30392570
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392570 25 mg 25mg
30394215 5 mg -
30402008 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30392570 Supplier Toronto Research Chemicals Supplier No. TRCH92450925MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C12 D3 H11 N2 O3
Molecular Weight (g/mol) 237.27
Synonym (3R-trans)-3-(Acetyloxy)-1-methyl-5-(3-pyridinyl)-2-pyrrolidinone-d3, (3'R,5'S)-3'-Hydroxy Cotinine-d3 Acetate, Hydroxycotinine-d3 Acetate
IUPAC Name [(3R,5S)-2-oxo-5-pyridin-3-yl-1-(trideuteriomethyl)pyrrolidin-3-yl] acetate
SMILES [2H]C([2H])([2H])N1[C@@H](C[C@@H](OC(=O)C)C1=O)c2cccnc2

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C12H14N2O3/c1-8(15)17-11-6-10(14(2)12(11)16)9-4-3-5-13-7-9/h3-5,7,10-11H,6H2,1-2H3/t10-,11+/m0/s1/i2D3
Formula Weight 237.119
Chemical Name or Material trans-3'-Hydroxy Cotinine-d3 Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.