missing translation for 'onlineSavingsMsg'
Learn More

trans-3'-Hydroxy Cotinine Acetate, TRC

Product Code. 30391848
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391848 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30391848 Supplier Toronto Research Chemicals Supplier No. TRCH9245085MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3'R,5'S)-3'-Hydroxycotinine Acetate
CAS 111034-55-0
Molecular Formula C12 H14 N2 O3
Quantity 5 mg
Synonym 2-Pyrrolidinone, 3-(acetyloxy)-1-methyl-5-(3-pyridinyl)-, (3R-trans)- (9CI), Hydroxycotinine acetate, trans-3'-Hydroxycotinine acetate, trans-3'-Hydroxy cotinine acetate, (3'R,5'S)-3'-Hydroxycotinine acetate, (3R,5S)-3-Acetoxy-1-methyl-5-(3-pyridinyl)-2-pyrrolidinone
InChI Formula InChI=1S/C12H14N2O3/c1-8(15)17-11-6-10(14(2)12(11)16)9-4-3-5-13-7-9/h3-5,7,10-11H,6H2,1-2H3/t10-,11+/m1/s1
SMILES CN1[C@H](C[C@H](OC(=O)C)C1=O)c2cccnc2
IUPAC Name [(3S,5R)-1-methyl-2-oxo-5-pyridin-3-ylpyrrolidin-3-yl] acetate
Molecular Weight (g/mol) 234.25
Formula Weight 234.1004
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.