missing translation for 'onlineSavingsMsg'
Learn More

trans-3-Hexene, 98%

Product Code. 11319576 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11319576 5 g Glass bottle 5g
10735863 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11319576 Supplier Thermo Scientific Acros Supplier No. 149910051
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13269-52-8 | C6H12 | 84.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13269-52-8
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
InChI Key ZQDPJFUHLCOCRG-AATRIKPKSA-N
Synonym trans-3-hexene, 3-hexene, e-3-hexene, 3-hexene, e, 3-hexene, 3e, unii-3co1lux1ar, e-hex-3-ene, 3co1lux1ar, alkenes, c6, cis-3-hexene
PubChem CID 638066
IUPAC Name (E)-hex-3-ene
SMILES CCC=CCC

Specifications

Melting Point -113.0°C
Density 0.6770g/mL
Boiling Point 67.0°C
Flash Point −12°C
Assay Percent Range 97.5% min
Packaging Glass bottle
Linear Formula C2H5CH=CHC2H5
Refractive Index 1.3935 to 1.3955
Quantity 5 g
Beilstein 01,II,192
Specific Gravity 0.677
Formula Weight 84.16
Percent Purity 98%
Chemical Name or Material trans-3-Hexene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • trans-3-Hexene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.