missing translation for 'onlineSavingsMsg'
Learn More

trans-3-Hexene, 98%

Product Code. 11378365
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378365 1 g 1g
11388365 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11378365 Supplier Thermo Scientific Alfa Aesar Supplier No. L10311.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13269-52-8 | C6H12 | 84.162 g/mol

Oligomers were prepared from the ozonolyses of cis- and trans-3-hexene and cis- and trans-2,5-dimethyl-3-hexene. Synthesis and characterization of aliphatic unsaturated polyesters is from trans-3-hexene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Oligomers were prepared from the ozonolyses of cis- and trans-3-hexene and cis- and trans-2,5-dimethyl-3-hexene. Synthesis and characterization of aliphatic unsaturated polyesters is from trans-3-hexene.

Solubility
Insoluble in water. Soluble in benzene, ligroin, ethanol, chloroform, ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13269-52-8
Molecular Formula C6H12
Molecular Weight (g/mol) 84.162
MDL Number MFCD00009386
InChI Key ZQDPJFUHLCOCRG-AATRIKPKSA-N
Synonym trans-3-hexene, 3-hexene, e-3-hexene, 3-hexene, e, 3-hexene, 3e, unii-3co1lux1ar, e-hex-3-ene, 3co1lux1ar, alkenes, c6, cis-3-hexene
PubChem CID 638066
IUPAC Name (E)-hex-3-ene
SMILES CCC=CCC

Specifications

Melting Point -113°C
Density 0.677
Boiling Point 66°C to 67°C
Flash Point −25°C (−13°F)
Odor Mild
Refractive Index 1.394
Quantity 1 g
UN Number UN3295
Beilstein 1718859
Solubility Information Insoluble in water. Soluble in benzene,ligroin,ethanol,chloroform,ether.
Formula Weight 84.16
Percent Purity 98%
Chemical Name or Material trans-3-Hexene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.