missing translation for 'onlineSavingsMsg'
Learn More

trans-3-Heptene, 97%

Product Code. 15470348
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15470348 25 g 25g
15450348 1 g 1g
15460348 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15470348 Supplier Thermo Scientific Alfa Aesar Supplier No. H53482.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14686-14-7 | C7H14 | 98.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14686-14-7
Molecular Formula C7H14
Molecular Weight (g/mol) 98.19
MDL Number MFCD00009475
InChI Key WZHKDGJSXCTSCK-FNORWQNLSA-N
Synonym trans-3-heptene, e-hept-3-ene, e-3-heptene, 3-heptene, e, 3-heptene, 3e, unii-o4br0a824i, hept-3-ene, cis-3-heptene, 3-heptene c,t, z-hept-3-ene
PubChem CID 5357259
SMILES CCC\C=C\CC

Specifications

Melting Point -137°C
Density 0.698
Boiling Point 94°C
Flash Point −1°C (30°F)
Refractive Index 1.403
Quantity 25 g
UN Number UN3295
IUPAC Name (E)-hept-3-ene
Formula Weight 98.19
Percent Purity 97%
Chemical Name or Material trans-3-Heptene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.