missing translation for 'onlineSavingsMsg'
Learn More

trans-3-(Boc-amino)cyclobutanecarboxylic acid, 97%, Thermo Scientific Chemicals

Product Code. 15419358
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15419358 1 g 1g
15447976 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15419358 Supplier Thermo Scientific Alfa Aesar Supplier No. H62902.03
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 939400-34-7 | Molecular Formula: C10H17NO4 | Molecular Weight: 215.249 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 939400-34-7
Molecular Formula C10H17NO4
Molecular Weight (g/mol) 215.249
MDL Number MFCD06657680
InChI Key KLCYDBAYYYVNFM-UHFFFAOYSA-N
Synonym cis-3-tert-butoxycarbonylamino cyclobutanecarboxylic acid, trans-3-tert-butoxycarbonylamino cyclobutanecarboxylic acid, 3-tert-butoxycarbonylamino cyclobutanecarboxylic acid, trans-3-tert-butoxycarbonyl amino cyclobutanecarboxylic acid, 3-boc-amino cyclobutanecarboxylic acid, 3-tert-butoxycarbonyl amino cyclobutanecarboxylic acid, cis-3-tert-butoxycarbonyl amino cyclobutanecarboxylic acid, cis-3-boc-amino cyclobutanecarboxylic acid, 3-tert-butoxy carbonyl amino cyclobutane-1-carboxylic acid, cis-3-boc-aminocyclobutanecarboxylic acid
PubChem CID 24720963
IUPAC Name 3-[(2-methylpropan-2-yl)oxycarbonylamino]cyclobutane-1-carboxylic acid
SMILES CC(C)(C)OC(=O)NC1CC(C1)C(=O)O

Specifications

Quantity 1 g
Formula Weight 215.24
Percent Purity 97%
Chemical Name or Material trans-3-(Boc-amino)cyclobutanecarboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.