missing translation for 'onlineSavingsMsg'
Learn More

trans-3-(4-Pyridyl)acrylic acid, 97%

Product Code. 11383077
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11383077 5 g 5g
11373077 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11383077 Supplier Thermo Scientific Alfa Aesar Supplier No. L13458.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 84228-93-3 | C8H7NO2 | 149.149 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84228-93-3
Molecular Formula C8H7NO2
Molecular Weight (g/mol) 149.149
MDL Number MFCD00040746
InChI Key SSAYTINUCCRGDR-OWOJBTEDSA-N
Synonym 3-4-pyridyl acrylic acid, trans-3-4-pyridyl acrylic acid, 3-pyridin-4-yl-acrylic acid, 4-pyridineacrylic acid, e-3-pyridin-4-yl acrylic acid, 3-pyridin-4-yl acrylic acid, 2e-3-pyridin-4-ylacrylic acid, 3-4-pyridine acrylic acid, 2e-3-pyridin-4-yl prop-2-enoic acid, e-3-pyridin-4-yl-acrylic acid
PubChem CID 736782
IUPAC Name (E)-3-pyridin-4-ylprop-2-enoic acid
SMILES C1=CN=CC=C1C=CC(=O)O

Specifications

Melting Point 276°C to 280°C
Quantity 5 g
Beilstein 1719967
Formula Weight 149.15
Percent Purity 97%
Chemical Name or Material trans-3-(4-Pyridyl)acrylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.