missing translation for 'onlineSavingsMsg'
Learn More

trans-3-(3-Pyridyl)acrylic acid, 99%

Product Code. 11457796
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
Unit Size:
250g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11457796 250 g 250g
11309559 5 g 5g
11319559 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11457796 Supplier Thermo Scientific Alfa Aesar Supplier No. A14747.30
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 19337-97-4 | C8H7NO2 | 149.149 g/mol

trans-3-(3-Pyridyl)acrylic acid is a useful synthetic intermediate. It is also used to prepare aminomethyl benzimidazoles, which acts as an inhibitor of gelatinase B. Further, it is used in the preparation of aminopyridines as antibacterial enoyl acyl carrier protein reductase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-3-(3-Pyridyl)acrylic acid is a useful synthetic intermediate. It is also used to prepare aminomethyl benzimidazoles, which acts as an inhibitor of gelatinase B. Further, it is used in the preparation of aminopyridines as antibacterial enoyl acyl carrier protein reductase inhibitors.

Notes
Incompatible with oxidizing agents, acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19337-97-4
Molecular Formula C8H7NO2
Molecular Weight (g/mol) 149.149
MDL Number MFCD00006410
InChI Key VUVORVXMOLQFMO-ONEGZZNKSA-N
Synonym 3-3-pyridyl acrylic acid, trans-3-3-pyridyl acrylic acid, e-3-pyridin-3-yl acrylic acid, 3-pyridylacrylic acid, 2e-3-pyridin-3-ylacrylic acid, 2e-3-pyridin-3-yl prop-2-enoic acid, 3-pyridin-3-yl acrylic acid, trans-3-3'-pyridyl acrylic acid, 3-3-pyridinyl acrylic acid, 3-pyridinyl-acrylic acid
PubChem CID 776396
IUPAC Name (E)-3-pyridin-3-ylprop-2-enoic acid
SMILES C1=CC(=CN=C1)C=CC(=O)O

Specifications

Melting Point 232°C to 235°C
Quantity 250 g
Beilstein 113290
Formula Weight 149.15
Percent Purity 99%
Chemical Name or Material trans-3-(3-Pyridyl)acrylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.