missing translation for 'onlineSavingsMsg'
Learn More

trans-3-(2-Chloro-3,3,3-trifluoro-1-propenyl)-2,2-dimethyl-cyclopropanecarboxylic Acid, TRC

Product Code. 30366302
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30366302 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30366302 Supplier Toronto Research Chemicals Supplier No. TRCC42201025MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material trans-3-(2-Chloro-3,3,3-trifluoro-1-propenyl)-2,2-dimethyl-cyclopropanecarboxylic Acid
CAS 72748-68-6
Molecular Formula C9 H10 Cl F3 O2
Quantity 25 mg
Synonym (1R,3S)-rel-3-(2-Chloro-3,3,3-trifluoro-1-propenyl)-2,2-dimethyl-cyclopropanecarboxylic Acid, trans-(±)-3-(2-Chloro-3,3,3-trifluoro-1-propenyl)-2,2-dimethyl-cyclopropanecarboxylic Acid
InChI Formula InChI=1S/C9H10ClF3O2/c1-8(2)4(6(8)7(14)15)3-5(10)9(11,12)13/h3-4,6H,1-2H3,(H,14,15)/b5-3-/t4-,6+/m1/s1
SMILES CC1(C)[C@H](\C=C(/Cl)\C(F)(F)F)[C@H]1C(=O)O
IUPAC Name (1R,3S)-3-[(Z)-2-chloro-3,3,3-trifluoroprop-1-enyl]-2,2-dimethylcyclopropane-1-carboxylic acid
Molecular Weight (g/mol) 242.623
Formula Weight 242.032
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.