missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Undecenal, 95%, remainder mainly cis-isomer, Thermo Scientific™

Product Code. 11382516
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11382516 5 g 5g
11392516 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11382516 Supplier Thermo Scientific Alfa Aesar Supplier No. L06437.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53448-07-0
Molecular Formula C11H20O
Molecular Weight (g/mol) 168.28
MDL Number MFCD00014680
InChI Key PANBRUWVURLWGY-MDZDMXLPSA-N
Synonym trans-2-undecenal, 2-undecenal, e-undec-2-enal, e-2-undecenal, 2-undecenal, 2e, 2-undecen-1-al, 2e-2-undecenal, trans-2-undecen-1-al, 2-undecenal, e, unii-fnp3s9mg30
PubChem CID 5283356
IUPAC Name (E)-undec-2-enal
SMILES CCCCCCCCC=CC=O

Specifications

Density 0.844
Boiling Point 115°C (10 mmHg)
Flash Point 104°C (219°F)
Refractive Index 1.457
Quantity 5 g
Beilstein 1762239
Sensitivity Air sensitive
Formula Weight 168.28
Percent Purity 95%
Assay remainder mainly cis-isomer
Chemical Name or Material trans-2-Undecenal
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.