missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Phenylcyclopropylamine hydrochloride, 97%

Product Code. 10634992 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10634992 250 mg Glass bottle 250mg
10624992 1 g Glass bottle 1g
10701735 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10634992 Supplier Thermo Scientific Acros Supplier No. 130472500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1986-47-6 | C9H11N·HCl | 169.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1986-47-6
Molecular Formula C9H11N·HCl
Molecular Weight (g/mol) 169.66
MDL Number MFCD00063602
InChI Key ZPEFMSTTZXJOTM-VTLYIQCISA-N
Synonym tranylcypromine hydrochloride, trans-2-phenylcyclopropanamine hydrochloride, trans-2-phenylcyclopropylamine hydrochloride, 1r,2s-2-phenyl-cyclopropylamine hydrochloride, 1r,2s-2-phenylcyclopropan-1-amine hydrochloride, tranylcypromine hcl, 1r,2s-2-phenylcyclopropanamine hydrochloride, trans-2-phenyl-cyclopropylamine hydrochloride, dsstox_cid_27761, dsstox_rid_82538
PubChem CID 12345947
IUPAC Name (1R,2R)-2-phenylcyclopropan-1-amine;hydrochloride
SMILES C1C(C1N)C2=CC=CC=C2.Cl

Specifications

Melting Point 162°C to 169°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Packaging Glass bottle
Linear Formula C6H5C3H4NH2·HCl
Quantity 250 mg
Beilstein 12, III, 2797
Merck Index 14, 9573
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol,ethyl acetate,diethyl ether
Formula Weight 169.66
Percent Purity 97%
Physical Form Chunks and/or Powder
Chemical Name or Material trans-2-Phenylcyclopropylamine hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • trans-2-Phenylcyclopropylamine hydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.