missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Pentenoic acid, 97%

Product Code. 10731341 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10731341 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10731341 Supplier Thermo Scientific Acros Supplier No. 279370250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13991-37-2 | C5H8O2 | 100.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 13991-37-2
Melting Point 9.0°C
Color Colorless to Yellow
Density 0.9900g/mL
Boiling Point 106.0°C (20.0 mmHg)
Flash Point 102°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C5H8O2
Linear Formula C2H5CH=CHCO2H
Refractive Index 1.4522
MDL Number MFCD00002704
Quantity 25 g
Beilstein 02, 426
Specific Gravity 0.99
Synonym trans-2-pentenoic acid, 2-pentenoic acid, e-pent-2-enoic acid, pent-2-enoic acid, 2e-pent-2-enoic acid, 2-pentenoic acid, 2e, beta-ethyl acrylic acid, c2h5ch=chcooh, pentenoic acid, unii-1rg66883cf
Solubility Information Solubility in water: 60g/L (20°C)
InChI Key YIYBQIKDCADOSF-ONEGZZNKSA-N
SMILES CC\C=C\C(O)=O
IUPAC Name (2E)-pent-2-enoic acid
Molecular Weight (g/mol) 100.12
PubChem CID 638122
ChEBI CHEBI:38366
Formula Weight 100.12
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material trans-2-Pentenoic acid, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • trans-2-Pentenoic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.