missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Octene 98%, Thermo Scientific™

Product Code. 10534594 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10534594 5 g Glass bottle 5g
10205010 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10534594 Supplier Thermo Scientific Acros Supplier No. 149990050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13389-42-9
Molecular Formula C8H16
Molecular Weight (g/mol) 112.22
MDL Number MFCD00009532
InChI Key ILPBINAXDRFYPL-HWKANZROSA-N
Synonym trans-2-octene, 2-octene, e-2-octene, e-oct-2-ene, 2-octene, e, 2-octene, 2e, unii-g13g1yr8yw, oct-2-ene, g13g1yr8yw, octene-2
PubChem CID 5364448
IUPAC Name (2E)-oct-2-ene
SMILES CCCCC\C=C\C

Specifications

Color Colorless
Density 0.7180g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3(CH2)4CH=CHCH3
Refractive Index 1.4120 to 1.4140 (20°C, 589nm)
Quantity 5 g
Formula Weight 112.21
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material trans-2-Octene, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.