missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Octene, 97%

Product Code. 11343266
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11343266

Brand: Thermo Scientific Alfa Aesar L12606.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13389-42-9 | C8H16 | 112.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13389-42-9
Molecular Formula C8H16
Molecular Weight (g/mol) 112.22
MDL Number MFCD00009532
InChI Key ILPBINAXDRFYPL-HWKANZROSA-N
Synonym trans-2-octene, 2-octene, e-2-octene, e-oct-2-ene, 2-octene, e, 2-octene, 2e, unii-g13g1yr8yw, oct-2-ene, g13g1yr8yw, octene-2
PubChem CID 5364448
IUPAC Name (E)-oct-2-ene
SMILES CCCCC\C=C\C

Specifications

Melting Point -89°C to -86°C
Density 0.718
Boiling Point 122°C to 123°C
Flash Point 14°C (57°F)
Refractive Index 1.413
Quantity 25 g
UN Number UN3295
Beilstein 1719497
Formula Weight 112.22
Percent Purity 97%
Chemical Name or Material trans-2-Octene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.