missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Nonenal, 97%

Product Code. 11489256
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11489256

Brand: Thermo Scientific Alfa Aesar A18108.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18829-56-6 | C9H16O | 140.226 g/mol

trans-2-Nonenal reacts with acetic acid hydrazide to produce acetic acid non-2-enylidene-hydrazide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-2-Nonenal reacts with acetic acid hydrazide to produce acetic acid non-2-enylidene-hydrazide.

Solubility
Soluble in Alcohol , and oils. Insoluble in water.

Notes
Air Sensitive. Incompatible with air, oxidizing agents and heat. Protect from heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18829-56-6
Molecular Formula C9H16O
Molecular Weight (g/mol) 140.226
MDL Number MFCD00007012
InChI Key BSAIUMLZVGUGKX-BQYQJAHWSA-N
Synonym trans-2-nonenal, 2-nonenal, e-2-nonenal, e-non-2-enal, 3-hexylacrolein, 2-nonenal, 2e, beta-hexylacrolein, 2-nonenal, e, heptylideneacetaldehyde, alpha-nonenyl aldehyde
PubChem CID 5283335
IUPAC Name (E)-non-2-enal
SMILES CCCCCCC=CC=O

Specifications

Density 0.846
Boiling Point 98°C to 100°C (12 mmHg)
Flash Point 84°C (184°F)
Refractive Index 1.4403
Quantity 5 g
Sensitivity Air sensitive
Merck Index 14,6676
Solubility Information Soluble in Alcohol ,and oils. Insoluble in water.
Formula Weight 140.22
Percent Purity 97%
Chemical Name or Material trans-2-Nonenal
Show More Show Less
compliance-icons
Product Identifier
  • trans-2-Nonenal
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : trans-2-Nonenal

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.