missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Nonenal, 95%, Thermo Scientific™

Product Code. 10636101 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
This item is not returnable. View return policy

Product Code. 10636101

Brand: Thermo Scientific Acros 298310250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 18829-56-6
Color Colorless to Yellow
Density 0.8400g/mL
Boiling Point 55°C to 60°C (0.3 mmHg)
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Molecular Formula C9H16O
Linear Formula CH3(CH2)5CH=CHCHO
Refractive Index 1.4520 to 1.456
MDL Number MFCD00007012
Quantity 25 mL
Merck Index 15, 6763
Specific Gravity 0.84
Synonym trans-2-nonenal, 2-nonenal, e-2-nonenal, e-non-2-enal, 3-hexylacrolein, 2-nonenal, 2e, beta-hexylacrolein, 2-nonenal, e, heptylideneacetaldehyde, alpha-nonenyl aldehyde
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,fixed oils
InChI Key BSAIUMLZVGUGKX-BQYQJAHWSA-N
SMILES CCCCCCC=CC=O
IUPAC Name (E)-non-2-enal
Molecular Weight (g/mol) 140.23
PubChem CID 5283335
Formula Weight 140.23
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material trans-2-Nonenal, 95%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.