missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Methyl-2-pentenoic acid, 97%, Thermo Scientific™

Catalog Number 15426858
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
16957-70-3
C6H10O2
114.144
MFCD00002655
JJYWRQLLQAKNAD-SNAWJCMRSA-N
2-methyl-2-pentenoic acid, trans-2-methyl-2-pentenoic acid, e-2-methylpent-2-enoic acid, e-2-methyl-2-pentenoic acid, 2-pentenoic acid, 2-methyl, 2-pentenoic acid, 2-methyl-, e, e-2-methylpent-2-en-1-oic acid, s-2-methylpentenoic acid, 2e-2-methylpent-2-enoic acid, 2-methylpent-2-en-1-oic acid
5365909
(E)-2-methylpent-2-enoic acid
CCC=C(C)C(=O)O
This item is not returnable. View return policy
16957-70-3
C6H10O2
114.144
MFCD00002655
JJYWRQLLQAKNAD-SNAWJCMRSA-N
2-methyl-2-pentenoic acid, trans-2-methyl-2-pentenoic acid, e-2-methylpent-2-enoic acid, e-2-methyl-2-pentenoic acid, 2-pentenoic acid, 2-methyl, 2-pentenoic acid, 2-methyl-, e, e-2-methylpent-2-en-1-oic acid, s-2-methylpentenoic acid, 2e-2-methylpent-2-enoic acid, 2-methylpent-2-en-1-oic acid
5365909
(E)-2-methylpent-2-enoic acid
CCC=C(C)C(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 26°C to 28°C
Density 0.979
Boiling Point 123°C to 125°C (30 mmHg)
Flash Point 108°C (226°F)
Linear Formula C2H5CH=C(CH3)CO2H
Refractive Index 1.46
Quantity 5 g
UN Number UN3261
Formula Weight 114.14
Percent Purity 97%
Chemical Name or Material trans-2-Methyl-2-pentenoic acid
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'