missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Hydroxymethyl-1-cyclohexylamine hydrochloride, 99+%, ACROS Organics™

Brand:  Acros Organics 265872500

Code : 29

Additional Details : CAS Number : 28250-45-5 Weight : 0.05025kg

Quantity 250mg
Packaging Glass bottle

 View more versions of this product

Product Code. 10319200

  • £83.20 / 250mg


Estimated Shipment
14-06-2018
Add to basket

Specifications

Chemical Identifiers

trans-2-Hydroxymethyl-1-cyclohexylamine hydrochloride
28250-45-5
99+%
165.66
99+%
≥99.0%
C7H15NO·HCl
MFCD00145424

Specifications

trans-2-Hydroxymethyl-1-cyclohexylamine hydrochloride
150° to 155°C
99+%
Authentic
Crystalline Powder
99+%
165.66
White to almost white
250mg
28250-45-5
Glass bottle
≥99.0%
C7H15NO·HCl
MFCD00145424
Videos
SDS