missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Hexenoic acid, 96%

Product Code. 11450084
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11450084 250 g 250g
11430084 10 g 10g
11440084 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11450084 Supplier Thermo Scientific Alfa Aesar Supplier No. A13571.30
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13419-69-7 | C6H9O2 | 113.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13419-69-7
Molecular Formula C6H9O2
Molecular Weight (g/mol) 113.14
MDL Number MFCD00002705
InChI Key NIONDZDPPYHYKY-SNAWJCMRSA-M
Synonym trans-2-hexenoic acid, 2-hexenoic acid, e-hex-2-enoic acid, hex-2-enoic acid, trans-hex-2-enoic acid, e-2-hexenoic acid, hexenoic acid, 2e-hex-2-enoic acid, 2-hexenoic acid, 2e, 2-hexenoic acid, e
PubChem CID 5282707
ChEBI CHEBI:87721
IUPAC Name (E)-hex-2-enoic acid
SMILES CCC\C=C\C([O-])=O

Specifications

Melting Point 30°C to 35°C
Boiling Point 217°C
Flash Point 125°C (257°F)
Quantity 250 g
UN Number UN3261
Beilstein 1720443
Formula Weight 114.14
Percent Purity 96%
Chemical Name or Material trans-2-Hexenoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.