Learn More
trans-2-Hexene, 99%
Description
trans-2-Hexene is used as a co monomer in the production of polyethylene. The use of trans-2-hexene, which is derived from 3-hexyne using sodium in liquid ammonia.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
trans-2-Hexene is used as a co monomer in the production of polyethylene. The use of trans-2-hexene, which is derived from 3-hexyne using sodium in liquid ammonia.
Solubility
Difficult to mix.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Melting Point | -99°C |
| Density | 0.674 |
| Boiling Point | 68°C to 69°C |
| Flash Point | −20°C (−4°F) |
| Odor | Mild |
| Refractive Index | 1.394 |
| Quantity | 10 g |
| UN Number | UN3295 |
| Beilstein | 1719020 |
| Solubility Information | Difficult to mix. |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.