missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Hexene, 98+%

Product Code. 10407170 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10407170 5 mL Glass bottle 5mL
10397120 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10407170 Supplier Thermo Scientific Acros Supplier No. 149900050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4050-45-7 | C6H12 | 84.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4050-45-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00009473
InChI Key RYPKRALMXUUNKS-HWKANZROSA-N
Synonym trans-2-hexene, e-2-hexene, 2-hexene, e, hexene, isomer, hexene, isomers, unii-2tz30ggg1a, 2e-hexene, e-hex-2-ene, 2tz30ggg1a, 2-hexene, 2e
PubChem CID 639661
IUPAC Name (E)-hex-2-ene
SMILES CCC\C=C\C

Specifications

Melting Point -98.5°C
Color Colorless
Density 0.6600g/mL
Boiling Point 68°C to 69°C
Flash Point -20°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH2CH2CH=CHCH3
Refractive Index 1.3920 to 1.394
Quantity 5 mL
Beilstein 01, I, 89
Specific Gravity 0.66
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in alcohol
Formula Weight 84.15
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material trans-2-Hexene, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • trans-2-Hexene
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.