missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Hexenal, TRC

Product Code. 30134432
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
Unit Size:
100g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134432 250 g 250g
30134336 100 g 100g
30008569 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134432 Supplier Toronto Research Chemicals Supplier No. TRCH294820250G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6728-26-3
Molecular Formula C6 H10 O
Molecular Weight (g/mol) 98.14
Synonym (E)-2-Hexenal, (2E)-2-Hexenal (ACI), 2-Hexenal, (E)- (8CI), Leaf aldehyde (6CI), (2E)-Hexenal, (E)-2-Hexen-1-al, (E)-2-Hexenal, (E)-Hex-2-enal, Green leaf aldehyde, trans-2-Hexen-1-al, trans-2-Hexenal
IUPAC Name (E)-hex-2-enal
SMILES CCC\C=C\C=O

Specifications

Quantity 250 g
InChI Formula InChI=1S/C6H10O/c1-2-3-4-5-6-7/h4-6H,2-3H2,1H3/b5-4+
Formula Weight 98.0732
Chemical Name or Material trans-2-Hexen-1-al
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.