missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Hexenal, 96%

Product Code. 11474503
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11474503 500 g 500g
11454503 25 g 25g
11464503 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11474503 Supplier Thermo Scientific Alfa Aesar Supplier No. A11513.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 6728-26-3 | C6H10O | 98.145 g/mol

trans-2-Hexenal is used in evaluation of quality of protected designation of origin virgin olive oils by headspace solid-phase microextraction-gas chromatography using flame ionization detection and multivariate analysis. It was used in the multicomponent method for the determination of low-molecular-weight carbonyl compounds in biological samples.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-2-Hexenal is used in evaluation of quality of protected designation of origin virgin olive oils by headspace solid-phase microextraction-gas chromatography using flame ionization detection and multivariate analysis. It was used in the multicomponent method for the determination of low-molecular-weight carbonyl compounds in biological samples.

Solubility
Not miscible in water.

Notes
Store at 4°C. Incompatible materials are bases, air, oxidizing agents, reducing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6728-26-3
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
MDL Number MFCD00007008
InChI Key MBDOYVRWFFCFHM-SNAWJCMRSA-N
Synonym trans-2-hexenal, 2-hexenal, leaf aldehyde, e-2-hexenal, e-hex-2-enal, 2-trans-hexenal, beta-propyl acrolein, hexylenic aldehyde, hex-2-en-1-al, 2-hexenal, e
PubChem CID 5281168
ChEBI CHEBI:28913
IUPAC Name (E)-hex-2-enal
SMILES CCCC=CC=O

Specifications

Density 0.848
Boiling Point 37°C to 38°C (8 mmHg)
Flash Point 38°C (100°F)
Odor Sweet
Refractive Index 1.447
Quantity 500 g
UN Number UN1988
Beilstein 1699684
Sensitivity Air sensitive
Solubility Information Not miscible in water.
Formula Weight 98.15
Percent Purity 96%
Chemical Name or Material trans-2-Hexenal
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.