missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Hexen-1-ol, 96%

Product Code. 10347380 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10347380 25 mL Glass bottle 25mL
10357380 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10347380 Supplier Thermo Scientific Acros Supplier No. 158140250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 928-95-0 | C6H12O | 100.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 928-95-0
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00002927
InChI Key ZCHHRLHTBGRGOT-SNAWJCMRSA-N
Synonym trans-2-hexen-1-ol, trans-2-hexenol, 2-hexenol, e-hex-2-en-1-ol, e-2-hexen-1-ol, 2-hexen-1-ol, 2-hexen-1-ol, 2e, 3-propylallyl alcohol, 2-hexen-1-ol, e, 2-hexen-1-ol, trans
PubChem CID 5318042
SMILES CCC\C=C\CO

Specifications

Color Colorless
Density 0.8440g/mL
Boiling Point 158.0°C to 160.0°C
Flash Point 54°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH2CH2CH=CHCH2OH
Refractive Index 1.4350 to 1.4410
Quantity 25 mL
Beilstein 01, II, 486
Specific Gravity 0.844
Solubility Information Solubility in water: slightly soluble.
IUPAC Name (E)-hex-2-en-1-ol
Formula Weight 100.16
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material trans-2-Hexen-1-ol, 96%
Show More Show Less
compliance-icons
Product Identifier
  • trans-2-Hexen-1-ol
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.