missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Decenal, 95%

Product Code. 10195840 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10195840 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10195840 Supplier Thermo Scientific Acros Supplier No. 298190250
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3913-81-3 | C10H18O | 154.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 3913-81-3
Color Colorless to Yellow
Density 0.8400g/mL
Boiling Point 78°C to 80°C (3.0 mmHg)
Flash Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Molecular Formula C10H18O
Linear Formula CH3(CH2)6CH=CHCHO
Refractive Index 1.4510 to 1.46
MDL Number MFCD00014679
Quantity 25 mL
Specific Gravity 0.84
Synonym trans-2-decenal, e-dec-2-enal, 2-decenal, decenal, e-2-decenal, 3-heptylacrolein, decenaldehyde, decylenic aldehyde, 2-decenal, 2e, 2-decenal, e
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,propylene glycol,essential,oil
InChI Key MMFCJPPRCYDLLZ-CMDGGOBGSA-N
SMILES CCCCCCC\C=C\C=O
IUPAC Name (2E)-dec-2-enal
Molecular Weight (g/mol) 154.25
PubChem CID 5283345
Formula Weight 154.25
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material trans-2-Decenal, 95%
Show More Show Less
compliance-icons compliance-icons
Produktidentifikator
  • trans-2-Decenal
Signalwort
  • Warning
Gefahrenkategorie
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Gefahrenhinweis
  • H400-Very toxic to aquatic life.
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Sicherheitshinweis
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.