missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Decenal, 95%

Product Code. p-7044326
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357625 5 g 5g
11367625 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357625 Supplier Thermo Scientific Alfa Aesar Supplier No. L05901.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3913-81-3 | C10H18O | 154.25 g/mol

trans-2-Decenal is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-2-Decenal is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Not miscible in water.

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3913-81-3
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00014679
InChI Key MMFCJPPRCYDLLZ-CMDGGOBGSA-N
Synonym trans-2-decenal, e-dec-2-enal, 2-decenal, decenal, e-2-decenal, 3-heptylacrolein, decenaldehyde, decylenic aldehyde, 2-decenal, 2e, 2-decenal, e
PubChem CID 5283345
SMILES CCCCCCC\C=C\C=O

Specifications

Density 0.84
Boiling Point 117°C (17 mmHg)
Flash Point 93°C (199°F)
Refractive Index 1.4565
Quantity 5 g
Beilstein 1704445
Solubility Information Not miscible in water.
IUPAC Name (E)-dec-2-enal
Formula Weight 154.25
Percent Purity 95%
Chemical Name or Material trans-2-Decenal
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.