missing translation for 'onlineSavingsMsg'
Learn More

trans-2,cis-6-Nonadienal, 90+% (major isomer), remainder mainly trans,trans-isomer

Product Code. 11493917
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493917 25 g 25g
11473917 1 g 1g
11483917 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11493917 Supplier Thermo Scientific Alfa Aesar Supplier No. A18356.14
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 557-48-2 | C9H14O | 138.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 557-48-2
Molecular Formula C9H14O
Molecular Weight (g/mol) 138.21
MDL Number MFCD00007009
InChI Key HZYHMHHBBBSGHB-ODYTWBPASA-N
Synonym trans-2,cis-6-nonadienal, violet leaf aldehyde, cucumber aldehyde, 2e,6z-nona-2,6-dienal, 2,6-nonadienal, 2e,6z, nona-2,6-dienal, e,z-2,6-nonadienal, 2e,6z-nonadienal, 2e,6z-nona-2,6-dien-1-al, 2-trans-6-cis-nonadienal
PubChem CID 643731
ChEBI CHEBI:7610
IUPAC Name (2E,6Z)-nona-2,6-dienal
SMILES CCC=CCCC=CC=O

Specifications

Density 0.866
Boiling Point 90°C (4 mmHg)
Flash Point 76°C (169°F)
Refractive Index 1.474
Quantity 25 g
Sensitivity Air Sensitive
Formula Weight 138.2
Percent Purity ≥90%
Assay remainder mainly trans,trans-isomer
Chemical Name or Material trans-2,cis-6-Nonadienal

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.