missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Aminocyclohexanol hydrochloride, 97+%

Product Code. 11466026
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11466026 100 g 100g
11446026 5 g 5g
11456026 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11466026 Supplier Thermo Scientific Alfa Aesar Supplier No. B22324.22
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5456-63-3 | C6H13ClNO | 150.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5456-63-3
Molecular Formula C6H13ClNO
Molecular Weight (g/mol) 150.63
MDL Number MFCD00001490,MFCD09259962,MFCD09259962,MFCD09259963,MFCD11618002,MFCD11618003,MFCD09259963
InChI Key SEBSRZIHRZRNAH-KGZKBUQUSA-N
PubChem CID 12228412
IUPAC Name (1R,2R)-2-aminocyclohexan-1-ol hydrochloridyl
SMILES [Cl].N[C@@H]1CCCC[C@H]1O

Specifications

Melting Point 172°C to 175°C
Quantity 100 g
Sensitivity Hygroscopic
Synonym trans-2-aminocyclohexanol hydrochloride, 1r,2r-2-aminocyclohexanol hydrochloride, 1r, 2r-2-aminocyclohexanol hydrochloride, 1r,2r-2-aminocyclohexanol hcl, 1r,2r-trans-2-aminocyclohexanol hydrochloride, trans-2-hydroxycyclohexylamine hydrochloride, 1r,2r-2-aminocyclohexan-1-ol hydrochloride, cyclohexanol, 2-amino-, hydrochloride, 1r,2r-rel, 1r,2r-2-aminocyclohexanolhydrochloride, 1r,2r-rel-2-aminocyclohexanol hydrochloride
Formula Weight 151.64
Percent Purity ≥98%
Chemical Name or Material trans-2-Aminocyclohexanol hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.