missing translation for 'onlineSavingsMsg'
Learn More

trans-2,6-Dimethylmorpholine, TRC

Product Code. 30383794
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383794 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30383794 Supplier Toronto Research Chemicals Supplier No. TRCD464990250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material trans-2,6-Dimethylmorpholine
CAS 6485-45-6
Molecular Formula C6H13NO
Quantity 250 mg
Synonym (2R,6R)-rel-2,6-Dimethylmorpholine
InChI Formula InChI=1S/C6H13NO/c1-5-3-7-4-6(2)8-5/h5-7H,3-4H2,1-2H3
SMILES C[C@H](CNC1)O[C@@H]1C
IUPAC Name (2R,6R)-2,6-dimethylmorpholine
Molecular Weight (g/mol) 115.17
Formula Weight 115.1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.