missing translation for 'onlineSavingsMsg'
Learn More

trans-2-(4-chlorophenyl)vinylboronic acid, 97%

Product Code. 10505490 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10505490 10 g Glass bottle 10g
10351720 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10505490 Supplier Thermo Scientific Acros Supplier No. 431340100
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 154230-29-2 | C8H8BClO2 | 182.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 154230-29-2
Molecular Formula C8H8BClO2
Molecular Weight (g/mol) 182.41
MDL Number MFCD02093767
InChI Key HWSDRAPTZRYXHN-AATRIKPKSA-N
Synonym trans-2-4-chlorophenyl vinylboronic acid, 4-chlorostyryl boronic acid, e-2-4-chlorophenyl ethenyl boronic acid, e-4-chlorostyrylboronic acid, boronic acid, 1e-2-4-chlorophenyl ethenyl, e-2-4-chlorophenyl ethenylboronic acid, trans-2-4-chlorophenyl ethenylboronic acid, rarechem al ba 0169, e-2-4-chlorophenyl vinyl boronic acid, 4-chlorostyrylboronic acid
PubChem CID 642694
IUPAC Name [(E)-2-(4-chlorophenyl)ethenyl]boronic acid
SMILES OB(O)\C=C\C1=CC=C(Cl)C=C1

Specifications

Melting Point 133.0°C to 134.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 10 g
Formula Weight 182.41
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material trans-2-(4-chlorophenyl)vinylboronic acid
compliance-icons
Product Identifier
  • trans-2-(4-Chlorophenyl)vinylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.