missing translation for 'onlineSavingsMsg'
Learn More

trans-(1R,2R)-N,N’-Bismethyl-1,2-cyclohexanediamine, TRC

Product Code. 30339973
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30339973 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30339973 Supplier Toronto Research Chemicals Supplier No. TRCB49480025G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material trans-(1R,2R)-N,N'-Bismethyl-1,2-cyclohexanediamine
CAS 67579-81-1
Molecular Formula C8 H18 N2
Quantity 25 g
Synonym trans-1,2-Bis(methylamino)cyclohexane, N,N’-Dimethyl-trans-1,2-cyclohexanediamine
InChI Formula InChI=1S/C8H18N2/c1-9-7-5-3-4-6-8(7)10-2/h7-10H,3-6H2,1-2H3/t7-,8-/m1/s1
SMILES CN[C@@H]1CCCC[C@H]1NC
IUPAC Name (1R,2R)-1-N,2-N-dimethylcyclohexane-1,2-diamine
Molecular Weight (g/mol) 142.24
Formula Weight 142.147
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.