missing translation for 'onlineSavingsMsg'
Learn More

trans-1,4-Dibromo-2-butene, 97%

Product Code. 10155363 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10155363 500 g Glass bottle 500g
10783603 25 g Glass bottle 25g
10374072 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10155363 Supplier Thermo Scientific Acros Supplier No. 406465000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 821-06-7 | C4H6Br2 | 213.9 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 821-06-7
Molecular Formula C4H6Br2
Molecular Weight (g/mol) 213.9
MDL Number MFCD00000249
InChI Key RMXLHIUHKIVPAB-OWOJBTEDSA-N
Synonym 1,4-dibromo-2-butene, trans-1,4-dibromo-2-butene, e-1,4-dibromobut-2-ene, e-1,4-dibromo-2-butene, 1,4-dibromobut-2-ene, trans-1,4-dibromobut-2-ene, 2-butene, 1,4-dibromo, 2-butene, 1,4-dibromo-, 2e, 2e-1,4-dibromobut-2-ene, 1,4-dibromo-2-butylene
PubChem CID 641245
IUPAC Name (E)-1,4-dibromobut-2-ene
SMILES C(C=CCBr)Br

Specifications

Melting Point 48°C to 53°C
Color White to Beige
Boiling Point 205°C
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula BrCH2CHCHCH2Br
Quantity 500 g
Solubility Information Solubility in water: slightly soluble. Other solubilities: slightly soluble in chloroform,soluble in ethanol,acetone
Formula Weight 213.9
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material trans-1, 4-Dibromo-2-butene, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • trans-1,4-Dibromo-2-butene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.