missing translation for 'onlineSavingsMsg'
Learn More

trans-1,4-Cyclohexane diisocyanate, 97%

Product Code. 11392827
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11392827 1 g 1g
11302837 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11392827 Supplier Thermo Scientific Alfa Aesar Supplier No. L15991.03
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 7517-76-2 | C8H10N2O2 | 166.18 g/mol

Mixed aromatic/alicyclic polyimides were prepared by polycondensation reactions of trans-1,4-cyclohexane diisocyanate CHDI with PMDA, BTDA, and 6FDA. The intermediate reactivity observed for CHDI, allows preparation of polyurethanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mixed aromatic/alicyclic polyimides were prepared by polycondensation reactions of trans-1,4-cyclohexane diisocyanate CHDI with PMDA, BTDA, and 6FDA. The intermediate reactivity observed for CHDI, allows preparation of polyurethanes.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture, water, alcohols, amines.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7517-76-2
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD00074787
InChI Key CDMDQYCEEKCBGR-UHFFFAOYSA-N
Synonym trans-1,4-diisocyanatocyclohexane, trans-1,4-cyclohexane diisocyanate, trans-1,4-cyclohexanediisocyanate, 1,4-cyclohexane diisocyanate, cyclohexane, 1,4-diisocyanato, cyclohexane, 1,4-diisocyanato-, trans, cyclohexane-1,4-diisocyanate, trans-1,4-cyclohexyleneisocyanate, 1,4-cyclohexyl di-isocyanate, cis-1,4-cyclohexylenediisocyanate
PubChem CID 17367
IUPAC Name 1,4-diisocyanatocyclohexane
SMILES C1CC(CCC1N=C=O)N=C=O

Specifications

Melting Point 59°C to 62°C
Boiling Point 145°C to 146°C (32 mmHg)
Odor Irritating
Quantity 1 g
UN Number UN2811
Beilstein 2804862
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water.
Formula Weight 166.18
Percent Purity 97%
Chemical Name or Material trans-1,4-Cyclohexane diisocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.