missing translation for 'onlineSavingsMsg'
Learn More

trans-1,3-Dichloropropene, TRC

Product Code. 30135080
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135080 1 g 1g
30134896 5 g 5g
30010576 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135080 Supplier Toronto Research Chemicals Supplier No. TRCD4345301G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10061-02-6
Molecular Formula C3 H4 Cl2
Molecular Weight (g/mol) 110.97
Synonym 1-Propene, 1,3-dichloro-, (E)-, Propene, 1,3-dichloro-, (E)- (8CI), (1E)-1,3-Dichloro-1-propene, (E)-1,3-Dichloro-1-propene, (E)-1,3-Dichloropropene, trans-1,3-Dichloro-1-propene, trans-1,3-Dichloropropene, trans-1,3-Dichloropropylene, trans-3-Chloroallyl chloride, trans-Telone
IUPAC Name (E)-1,3-dichloroprop-1-ene
SMILES ClC\C=C\Cl

Specifications

Quantity 1 g
InChI Formula InChI=1S/C3H4Cl2/c4-2-1-3-5/h1-2H,3H2/b2-1+
Formula Weight 109.969
Chemical Name or Material trans-1,3-Dichloropropene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.