missing translation for 'onlineSavingsMsg'
Learn More

trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate, 98%

Product Code. 11499987
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11499987 100 g 100g
11489987 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11499987 Supplier Thermo Scientific Alfa Aesar Supplier No. B22928.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate, 98%, C14H24N2O9, CAS Number-125572-95-4 | CAS: 125572-95-4 | C14H20N2O8 | 344.32 g/mol

trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate is an analytical reagent and a complexating agent for sodium. It acts as a ligand to prepare lanthanide shift reagents. It is involved in the separation and determination of iron(III)-dimethyldithiocarbamate, manganese(II)-ethylenebisdithiocarbamate and zinc(II)-ethylenebisdithiocarbamate by sensitive capillary electrophoretic ultraviolet method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-1,2-Diaminocyclohexane-N,N,N′,N′-tetraacetic acid monohydrate is an analytical reagent and a complexating agent for sodium. It acts as a ligand to prepare lanthanide shift reagents. It is involved in the separation and determination of iron(III)-dimethyldithiocarbamate, manganese(II)-ethylenebisdithiocarbamate and zinc(II)-ethylenebisdithiocarbamate by sensitive capillary electrophoretic ultraviolet method.

Solubility
Slightly soluble in water. Soluble in 1N sodium hydroxide and alkali solutions. Insoluble in most common organic solvents.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 125572-95-4
Molecular Formula C14H20N2O8
Molecular Weight (g/mol) 344.32
MDL Number MFCD00149243,MFCD00066429,MFCD00003845
InChI Key FCKYPQBAHLOOJQ-NXEZZACHSA-L
Synonym 2,2',2,2'-trans-cyclohexane-1,2-diylbis azanetriyl tetraacetic acid hydrate, dcyta, trans-1,2-diaminocyclohexane-n,n,n',n'-tetraacetic acid monohydrate, 1,2-cyclohexanedinitrilotetraacetic acid, 2,2',2,2'-1r,2r-rel-cyclohexane-1,2-diylbis azanetriyl tetraacetic acid hydrate, trans-1,2-cyclohexanediaminetetraacetic acid monohydrate, cdta monohydrate, ctda monohydrate, chel-cd, chel r-cd
PubChem CID 2723844
IUPAC Name 2-[[(1R,2R)-2-[bis(carboxymethyl)amino]cyclohexyl]-(carboxymethyl)amino]acetic acid;hydrate
SMILES [O-]C(=O)C[NH+](CC([O-])=O)[C@@H]1CCCC[C@H]1[NH+](CC([O-])=O)CC([O-])=O

Specifications

Melting Point ∼220°C (decomposition)
Quantity 100 g
Beilstein 2226756
Solubility Information Slightly soluble in water. Soluble in 1N sodium hydroxide and alkali solutions. Insoluble in most common organic solvents.
Formula Weight 364.35 (346.34 Anhydrous)
Percent Purity 98%
Chemical Name or Material trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.