missing translation for 'onlineSavingsMsg'
Learn More

trans-1,2-Cyclohexanediol, 98%

Product Code. 11317136
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11317136

Brand: Thermo Scientific Alfa Aesar L07088.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1460-57-7 | C6H12O2 | 116.16 g/mol

trans-1,2-Cyclohexanediol is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-1,2-Cyclohexanediol is used as an organic chemical synthesis intermediate.

Solubility
Soluble in chloroform and methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1460-57-7
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00063611
InChI Key PFURGBBHAOXLIO-PHDIDXHHSA-N
Synonym trans-1,2-cyclohexanediol, trans-cyclohexane-1,2-diol, 1r,2r-cyclohexane-1,2-diol, 1r,2r-trans-1,2-cyclohexanediol, 1,2-cyclohexanediol, 1r,2r-rel, trans-1,2-dihydroxycyclohexane, 1,2-cyclohexanediol, trans, l6tj aq bq&&trans, trans-1,2-cyclohexandiol, 1r,2r-1,2-cyclohexanediol
PubChem CID 92886
ChEBI CHEBI:16931
IUPAC Name (1R,2R)-cyclohexane-1,2-diol
SMILES O[C@@H]1CCCC[C@H]1O

Specifications

Melting Point 101°C to 106°C
Boiling Point 231°C to 233°C
Flash Point 134°C (273°F)
Quantity 100 g
Beilstein 3193810
Solubility Information Soluble in chloroform and methanol.
Formula Weight 116.16
Percent Purity 98%
Chemical Name or Material trans-1,2-Cyclohexanediol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.