missing translation for 'onlineSavingsMsg'
Learn More

trans-1,2-Cyclohexanediol, 98%

Product Code. 11307136
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11307136 25 g 25g
11317136 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11307136 Supplier Thermo Scientific Alfa Aesar Supplier No. L07088.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1460-57-7 | C6H12O2 | 116.16 g/mol

trans-1,2-Cyclohexanediol is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-1,2-Cyclohexanediol is used as an organic chemical synthesis intermediate.

Solubility
Soluble in chloroform and methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1460-57-7
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00063611
InChI Key PFURGBBHAOXLIO-PHDIDXHHSA-N
Synonym trans-1,2-cyclohexanediol, trans-cyclohexane-1,2-diol, 1r,2r-cyclohexane-1,2-diol, 1r,2r-trans-1,2-cyclohexanediol, 1,2-cyclohexanediol, 1r,2r-rel, trans-1,2-dihydroxycyclohexane, 1,2-cyclohexanediol, trans, l6tj aq bq&&trans, trans-1,2-cyclohexandiol, 1r,2r-1,2-cyclohexanediol
PubChem CID 92886
ChEBI CHEBI:16931
IUPAC Name (1R,2R)-cyclohexane-1,2-diol
SMILES O[C@@H]1CCCC[C@H]1O

Specifications

Melting Point 101°C to 106°C
Boiling Point 231°C to 233°C
Flash Point 134°C (273°F)
Quantity 25 g
Beilstein 3193810
Solubility Information Soluble in chloroform and methanol.
Formula Weight 116.16
Percent Purity 98%
Chemical Name or Material trans-1,2-Cyclohexanediol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.