missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-1,2-Cyclohexanedicarboxylic Acid, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | trans-1,2-Cyclohexanedicarboxylic Acid |
| CAS | 2305-32-0 |
| Molecular Formula | C8H12O4 |
| Quantity | 25 g |
| Synonym | (1R,2R)-rel-1,2-Cyclohexanedicarboxylic Acid, (±)-trans-1,2-Cyclohexanedicarboxylic Acid, NSC 31593 |
| InChI Formula | InChI=1S/C8H12O4/c9-7(10)5-3-1-2-4-6(5)8(11)12/h5-6H,1-4H2,(H,9,10)(H,11,12)/t5-,6-/m1/s1 |
| SMILES | O=C(O)[C@H]1[C@H](C(O)=O)CCCC1 |
| IUPAC Name | (1R,2R)-cyclohexane-1,2-dicarboxylic acid |
| Molecular Weight (g/mol) | 172.18 |
| Formula Weight | 172.0736 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction