missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tranilast, MedChemExpress
Description
Tranilast (MK-341) acts as an anti-atopic agent. Tranilast suppresses production of prostaglandin D2 (PGD2, IC50= 0.1 mM). Tranilast sodium exhibits anti-inflammatory and immunomodulatory effects. Tranilast sodium antagonizes angiotensin II and inhibits its biological effects in vascular smooth muscle cells.
Specifications
Specifications
| Chemical Name or Material | Tranilast |
| CAS | 53902-12-8 |
| Color | Yellowish Green |
| Purity Grade Notes | Research |
| Molecular Formula | C18H17NO5 |
| Quantity | 10 mg |
| Synonym | MK-341 SB 252218 |
| Solubility Information | DMSO : 50 mg/mL (152.75 mM; Need ultrasonic) |
| SMILES | O=C(O)C1=CC=CC=C1NC(/C=C/C2=CC=C(OC)C(OC)=C2)=O |
| Molecular Weight (g/mol) | 327.33 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?