missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tranilast, ACROS Organics™

Product Code. 17392481 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17392481 1 g 1g
17302491 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17392481 Supplier Thermo Scientific™ Supplier No. 468610010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tranilast
CAS 53902-12-8
Quantity 1 g
Solubility insoluble in water
Merck Index 9646
Melting Point 166.2°C to 168.2°C
Molecular Formula C18 H17 N O5
Formula Weight 327.33
Physical Form Crystalline Powder
Color White to Light-yellow to Green
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.