missing translation for 'onlineSavingsMsg'
Learn More

Tranexamic-d2 Acid, TRC

Product Code. 30454571
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30454571

Brand: Toronto Research Chemicals TRCT71450810MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tranexamic Acid D2 (aminomethyl-D2)
CAS 2714435-89-7
Molecular Formula C8H13D2NO2
Quantity 10 mg
Synonym trans-4-(Aminomethyl)cyclohexanecarboxylic-d2 Acid, AMCHA-d2, Amchafibrin-d2, Amikapron-d2, Anvitoff-d2, Bay 3517-d2, CL 65336-d2, Cyclocapron-d2, Cyclokapron-d2, Cyklokapron-d2, DV 79-d2, Exacyl-d2, HAKU-d2, Haematrix-d2, TAMCHA-d2, Tranol-d2, Transamin-d2, Transamin S-d2, Transcam-d2, Ugurol-d2, Xamig-d2, Xed-d2, Zataranax-d2, t-AMCHA-d2, trans-Amcha-d2, trans-p-(Aminomethyl)cyclohexanecarboxylic-d2 Acid
InChI Formula InChI=1S/C8H15NO2/c9-5-6-1-3-7(4-2-6)8(10)11/h6-7H,1-5,9H2,(H,10,11)/t6-,7-/i5D2
SMILES NC([2H])([2H])[C@@H]1CC[C@@H](C(O)=O)CC1
IUPAC Name (1r,4r)-4-(aminomethyl-d2)cyclohexane-1-carboxylic acid
Molecular Weight (g/mol) 159.22
Formula Weight 159.12
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.