missing translation for 'onlineSavingsMsg'
Learn More

Tranexamic Acid, TRC

Product Code. 17961575
Change view
Click to view available options
Quantity:
50 mg
1 g
10 g
Unit Size:
10g
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17961575 10 g 10g
17942985 1 g 1g
17921685 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17961575 Supplier Toronto Research Chemicals Supplier No. TRCT71450510G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1197-18-8
Molecular Formula C8 H15 N O2
Molecular Weight (g/mol) 157.21
Synonym Cyclohexanecarboxylic acid, 4-(aminomethyl)-, trans-, trans-4-(Aminomethyl)cyclohexanecarboxylic acid, 4-trans-Aminomethylcyclohexanecarboxylic acid, AMCA, AMCHA, Amchafibrin, Amikapron, Anvitoff, Bay 3517, CL 65336, Cyclocapron, Cyclokapron, Cyklokapron, DV 79, Exacyl, HAKU, Haematrix, Lysteda, TAMCHA, Tranexamic acid, Tranol, Transamin, Transamin G, Transamin S, Transcam, Ugurol, Xamig, Xed, Zataranax, t-AMCHA, trans-4-(Aminomethyl)-1-cyclohexanecarboxylic acid, trans-Amcha, trans-Tranexamic acid, trans-p-(Aminomethyl)cyclohexanecarboxylic acid
IUPAC Name 4-(aminomethyl)cyclohexane-1-carboxylic acid
SMILES NC[C@@H]1CC[C@H](CC1)C(=O)O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C8H15NO2/c9-5-6-1-3-7(4-2-6)8(10)11/h6-7H,1-5,9H2,(H,10,11)/t6-,7-
Formula Weight 157.1103
Percent Purity >95%
Chemical Name or Material Tranexamic acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.