missing translation for 'onlineSavingsMsg'
Learn More

Tranexamic acid, MedChemExpress

Product Code. 16470788
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16470788 5 g 5g
16428368 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16470788 Supplier Medchem Express Supplier No. HYB0149
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Tranexamic acid (Transamin) is an antifibrinolytic for blocking lysine-binding sites of plasmin and elastase-derived plasminogen fragments with IC50 of 5 mM.

Tranexamic acid (Transamin) is an antifibrinolytic for blocking lysine-binding sites of plasmin and elastase-derived plasminogen fragments with IC50 of 5 mM.

Specifications

Chemical Name or Material Tranexamic acid
Physical Form Solid
CAS 1197-18-8
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C8H15NO2
For Use With (Application) COVID-19-immunoregulation
Solubility Information H2O : 110 mg/mL (699.70 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble)
SMILES O=C([C@H]1CC[C@H](CN)CC1)O
Molecular Weight (g/mol) 157.21
Formula Weight 157.21
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.