missing translation for 'onlineSavingsMsg'
Learn More

Tranexamic acid, 98+%

Product Code. 10762461 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass Bottle
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10762461 50 g Glass Bottle 50g
10277453 250 g Glass Bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10762461 Supplier Thermo Scientific Acros Supplier No. 228040500
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1197-18-8 | C8H15NO2 | 157.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1197-18-8
Molecular Formula C8H15NO2
Molecular Weight (g/mol) 157.21
MDL Number MFCD00001466
InChI Key GYDJEQRTZSCIOI-UHFFFAOYSA-N
Synonym tranexamic acid, cyklokapron, tranexamsaeure, trans amcha, tranhexamic acid, trans-4-aminomethyl cyclohexanecarboxylic acid, transamin, amstat, tamcha, amikapron
PubChem CID 5526
IUPAC Name 4-(aminomethyl)cyclohexane-1-carboxylic acid
SMILES C1CC(CCC1CN)C(=O)O

Specifications

Melting Point >300°C
pH 6.5 to 8 (5% soln.)
Color White
Infrared Spectrum Authentic
Assay Percent Range 98+%
Loss on Drying 1% max.
Packaging Glass Bottle
Linear Formula H2NCH2C6H10CO2H
Merck Index 15, 9730
Solubility Information Solubility in water: 166g/L. Other solubilities: slightly soluble in alcohol and ether,practically insoluble in most organic solvents
Quantity 50 g
Formula Weight 157.21
Percent Purity ≥98%
Physical Form Crystalline Powder
Chemical Name or Material Tranexamic acid
Show More Show Less
compliance-icons
Product Identifier
  • trans-4-(Aminomethyl)cyclohexanecarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.