missing translation for 'onlineSavingsMsg'
Learn More

Tramiprosate, MedChemExpress

Product Code. 16458707
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16458707 5 g 5g
16401429 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16458707 Supplier Medchem Express Supplier No. HY14602
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Tramiprosate (Homotaurine), an orally active and brain-penetrant natural amino acid found in various species of red marine algae. Tramiprosate binds to soluble Aβ and maintains Aβ in a non-fibrillar form. Tramiprosate is also a GABA analog and possess neuroprotection, anticonvulsion and antihypertension effects.

Tramiprosate (Homotaurine), an orally active and brain-penetrant natural amino acid found in various species of red marine algae. Tramiprosate binds to soluble Aβ and maintains Aβ in a non-fibrillar form. Tramiprosate is also a GABA analog and possess neuroprotection, anticonvulsion and antihypertension effects.

Specifications

Chemical Name or Material Tramiprosate
CAS 3687-18-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C3H9NO3S
Quantity 5 g
Synonym Homotaurine 3-Amino-1-propanesulfonic acid
Solubility Information H2O : 20 mg/mL (143.71 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (7.19 mM; Need ultrasonic)
SMILES NCCCS(=O)(O)=O
Molecular Weight (g/mol) 139.17
Formula Weight 139.17
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.