missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Trametinib
Description
Trametinib is a potent and selective MEK1 and MEK2 inhibitor (IC50 = 0.92 and 1.8 nM, respectively, in cell-free assays). In vitro, it inhibits the growth of human colorectal cancer cell lines expressing either B-RAF or K-RAF (IC50 values range from 0.48 to 36 nM). In vivo, it suppresses tumor growth of HT-29 and COLO205 xenografts in nude mice. Trametinib induces apoptosis. This compound is orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | N-[3-[3-Cyclopropyl-5-[(2-fluoro-4-iodophenyl)amino]-3,4,6,7-tetrahydro-6,8-dimethyl-2,4,7-trioxopyrido[4,3-d]pyrimidin-1(2 H)-yl]phenyl]acetamide |
| CAS | 871700-17-3 |
| Quantity | 10 mg |
| Target | MEK Inhibitors |
| Molecular Formula | C26H23FIN5O4 |
| Purity | 0.98 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?